Please login to your online account to display your discounted pricing

4-(Thien-2-ylmethyl)benzaldehyde, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Thien-2-ylmethyl)benzaldehyde
Assay Percent Range 95%
Molecular Formula C12H10OS
Formula Weight 202.28
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

CC70604DE

  SDS
fisher scientific company
CC70604DE
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC70604CB

  SDS
fisher scientific company
CC70604CB
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC70604DA

  SDS
fisher scientific company
CC70604DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.28
Quantity 5g
Assay Percent Range 95%
Molecular Formula C12H10OS
Chemical Name or Material 4-(Thien-2-ylmethyl)benzaldehyde
CAS 1007847-72-4