Please login to your online account to display your discounted pricing

4-(Thien-2-ylmethyl)benzaldehyde, 95%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC70604DE

 View more versions of this product

Catalog No. CC70604DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 202.28
Quantity 5g
Molecular Formula C12H10OS
Assay Percent Range 95%
Chemical Name or Material 4-(Thien-2-ylmethyl)benzaldehyde
CAS 1007847-72-4

Specifications

Chemical Name or Material 4-(Thien-2-ylmethyl)benzaldehyde
Assay Percent Range 95%
Molecular Formula C12H10OS
Formula Weight 202.28