missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Tetrazol-5-yl)phenylboronic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445570050
Specifications
4-(Tetrazol-5-yl)phenylboronic acid | |
179942-55-3 | |
C7H7BN4O2 | |
DXUPJOQUAAVAGV-UHFFFAOYSA-N | |
[4-(2H-tetrazol-5-yl)phenyl]boronic acid | |
46737995 | |
97% |
5g | |
95% min. (HPLC) | |
4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid | |
B(C1=CC=C(C=C1)C2=NNN=N2)(O)O | |
189.97 | |
189.97 |
Chemical Identifiers
179942-55-3 | |
189.97 | |
4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid | |
[4-(2H-tetrazol-5-yl)phenyl]boronic acid |
C7H7BN4O2 | |
DXUPJOQUAAVAGV-UHFFFAOYSA-N | |
46737995 | |
B(C1=CC=C(C=C1)C2=NNN=N2)(O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning