missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Tetrahydropyran-4-yloxy)aniline, 97%, Maybridge™

Chemical Identifiers

CAS 917483-71-7
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD09064968
InChI Key CCIFXNITZHWCRR-UHFFFAOYSA-N
Synonym 4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenylamine, 4-oxan-4-yl oxy aniline, 4-oxan-4-yloxy aniline, 4-tetrahydro-2h-pyran-4-yl oxy aniline, 4-tetrahydro-2h-pyran-4-yloxy aniline, 4-tetrahydro-pyran-4-yloxy-phenylamine, 4-tetrahydropyran-4-yloxy aniline, 4-tetrahydropyran-4-yloxy phenylamine
PubChem CID 24229526
IUPAC Name 4-(oxan-4-yloxy)aniline
SMILES C1COCCC1OC2=CC=C(C=C2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC52414CB
SDS View Documents
Fisher Scientific Company
CC52414CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

917483-71-7
193.246
CCIFXNITZHWCRR-UHFFFAOYSA-N
24229526
C1COCCC1OC2=CC=C(C=C2)N
C11H15NO2
MFCD09064968
4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenylamine, 4-oxan-4-yl oxy aniline, 4-oxan-4-yloxy aniline, 4-tetrahydro-2h-pyran-4-yl oxy aniline, 4-tetrahydro-2h-pyran-4-yloxy aniline, 4-tetrahydro-pyran-4-yloxy-phenylamine, 4-tetrahydropyran-4-yloxy aniline, 4-tetrahydropyran-4-yloxy phenylamine
4-(oxan-4-yloxy)aniline

Specifications

Amber Glass Bottle
917483-71-7
100.0
C11H15NO2
4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenylamine, 4-oxan-4-yl oxy aniline, 4-oxan-4-yloxy aniline, 4-tetrahydro-2h-pyran-4-yl oxy aniline, 4-tetrahydro-2h-pyran-4-yloxy aniline, 4-tetrahydro-pyran-4-yloxy-phenylamine, 4-tetrahydropyran-4-yloxy aniline, 4-tetrahydropyran-4-yloxy phenylamine
C1COCCC1OC2=CC=C(C=C2)N
193.246
193.24
4-(Tetrahydropyran-4-yloxy)aniline
250mg
96.5
97%
MFCD09064968
CCIFXNITZHWCRR-UHFFFAOYSA-N
4-(oxan-4-yloxy)aniline
24229526
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS