Please login to your online account to display your discounted pricing

α,α,α,4-Tetrafluoro-m-toluidine, 99%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  203280050

 View more versions of this product

Catalog No. AC203280050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 179.12
Quantity 5mL
Color Clear yellow to brown-red
Density 1.3900g/mL
CAS Max % 100.0
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7 H5 F4 N
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
MDL Number MFCD00007834
Chemical Name or Material α,α,α,4-Tetrafluoro-m-toluidine
CAS Min % 98.5
CAS 2357-47-3
Linear Formula FC6H3(CF3)NH2
Packaging Glass bottle
Storage Information Product darkens on storage

Specifications

Chemical Name or Material α,α,α,4-Tetrafluoro-m-toluidine
CAS 2357-47-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7 H5 F4 N
Linear Formula FC6H3(CF3)NH2
Formula Weight 179.12
MDL Number MFCD00007834
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents