Learn More
4-tert-Pentylphenol, 99%, Thermo Scientific Chemicals
$119.53 - $119.53
Chemical Identifiers
CAS | 80-46-6 |
---|---|
Molecular Formula | C11H16O |
Molecular Weight (g/mol) | 164.248 |
MDL Number | MFCD00002369 |
InChI Key | NRZWYNLTFLDQQX-UHFFFAOYSA-N |
Synonym | 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol |
PubChem CID | 6643 |
ChEBI | CHEBI:35096 |
IUPAC Name | 4-(2-methylbutan-2-yl)phenol |
SMILES | CCC(C)(C)C1=CC=C(C=C1)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2093930
|
Thermo Scientific Chemicals
B2093930 |
250 g |
Each for $119.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
80-46-6 | |
164.248 | |
NRZWYNLTFLDQQX-UHFFFAOYSA-N | |
6643 | |
4-(2-methylbutan-2-yl)phenol |
C11H16O | |
MFCD00002369 | |
4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol | |
CHEBI:35096 | |
CCC(C)(C)C1=CC=C(C=C1)O |
Specifications
80-46-6 | |
0.962 | |
C11H16O | |
250 g | |
14,7142 | |
NRZWYNLTFLDQQX-UHFFFAOYSA-N | |
4-(2-methylbutan-2-yl)phenol | |
6643 | |
164.25 | |
4-tert-Pentylphenol |
88°C to 89°C | |
254°C to 256°C | |
MFCD00002369 | |
UN2430 | |
4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol | |
CCC(C)(C)C1=CC=C(C=C1)O | |
164.248 | |
CHEBI:35096 | |
99% |
Safety and Handling
GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber : 201-280-9
RTECSNumber : SM6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only