Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Pentylcyclohexanone, 97%

$44.00 - $504.77

Chemical Identifiers

Chemical Name or Material 4-tert-Pentylcyclohexanone
CAS 16587-71-6
Molecular Formula C11H20O
Formula Weight 168.28
MDL Number MFCD00013798
Beilstein 2240475
Synonym 4-tert-Amylcyclohexanone; 4-(1,1-Dimethylpropyl)cyclohexanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA128320B

  SDS
alfa aesar™
A128320B
1000g Each for $504.77

AAA1283218

  SDS
alfa aesar™
A1283218
50g Each for $44.00
Description & Specifications

Specifications

Formula Weight 168.28
Synonym 4-tert-Amylcyclohexanone; 4-(1,1-Dimethylpropyl)cyclohexanone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-tert-Pentylcyclohexanone
TSCA TSCA
Assay 97%
Molecular Formula C11H20O
MDL Number MFCD00013798
CAS 16587-71-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1000g
RTECS Number GW1960000
Beilstein 2240475
Boiling Point 124° to 125°C (16mmHg)

Applications
It finds its application as a masking agent and a perfuming agent in cosmetic industry.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.