Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Pentylbenzenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-tert-Pentylbenzenesulfonyl chloride
CAS 169677-20-7
Molecular Formula C11H15ClO2S
Formula Weight 246.76
MDL Number MFCD00041509
UN Number UN3261
Beilstein 2102813
Synonym 4-tert-Amylbenzenesulfonyl chloride; 4-(1,1-Dimethylpropyl)benzenesulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1089403

  SDS
alfa aesar™
L1089403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1089406

  SDS
alfa aesar™
L1089406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.76
CAS 169677-20-7
Molecular Formula C11H15ClO2S
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 4-tert-Amylbenzenesulfonyl chloride; 4-(1,1-Dimethylpropyl)benzenesulfonyl chloride
MDL Number MFCD00041509
Assay 97%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN3261
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 44° to 49°C
Beilstein 2102813
Quantity 1g
Health Hazard 1 Danger
Chemical Name or Material 4-tert-Pentylbenzenesulfonyl chloride
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.