missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Pentylbenzenesulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ L1089406

 View more versions of this product

Catalog No. AAL1089406



Specifications

Specifications

4-tert-Pentylbenzenesulfonyl chloride
5g
C11H15ClO2S
UN3261
4-tert-pentylbenzenesulfonyl chloride, 4-tert-amylbenzenesulphonyl chloride, 4-2-methylbutan-2-yl benzene-1-sulfonyl chloride, 4-tert-amylbenzenesulfonyl chloride, 4-tert-amyl-benzenesulphonyl chloride, 4-1,1-dimethylpropyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl benzenesulfonyl chloride, acmc-1bzun, 4-t-pentylbenzenesulfonyl chloride
CCC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl
246.749
246.76
44°C to 49°C
169677-20-7
MFCD00041509
2102813
WLBQFZZPRWQTGR-UHFFFAOYSA-N
4-(2-methylbutan-2-yl)benzenesulfonyl chloride
22281976
97%

Chemical Identifiers

169677-20-7
246.749
WLBQFZZPRWQTGR-UHFFFAOYSA-N
22281976
CCC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl
C11H15ClO2S
MFCD00041509
4-tert-pentylbenzenesulfonyl chloride, 4-tert-amylbenzenesulphonyl chloride, 4-2-methylbutan-2-yl benzene-1-sulfonyl chloride, 4-tert-amylbenzenesulfonyl chloride, 4-tert-amyl-benzenesulphonyl chloride, 4-1,1-dimethylpropyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl benzenesulfonyl chloride, acmc-1bzun, 4-t-pentylbenzenesulfonyl chloride
4-(2-methylbutan-2-yl)benzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents