Learn More
4-tert-Butyltoluene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 296320010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-tert-Butyltoluene | |
95.0 | |
-54.0°C | |
0.8612g/mL | |
54°C | |
95% min. (GC) | |
C11H16 | |
(CH3)3CC6H4CH3 | |
1 L | |
0.8612 | |
Solubility in water: 0.06 wt% (20°C). Other solubilities: miscible with isopropanol,toluene and cyclohexane,soluble in nearly all organic solvents | |
CC1=CC=C(C=C1)C(C)(C)C | |
148.25 | |
1.25 mPa.s (25°C) | |
96% |
98-51-1 | |
100.0 | |
Colorless to Yellow | |
189.0°C to 192.0°C | |
Authentic | |
Glass bottle | |
1.4910 to 1.4930 | |
MFCD00008837 | |
05, 439 | |
4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene | |
QCWXDVFBZVHKLV-UHFFFAOYSA-N | |
1-tert-butyl-4-methylbenzene | |
7390 | |
148.25 | |
Liquid |
Chemical Identifiers
98-51-1 | |
148.25 | |
QCWXDVFBZVHKLV-UHFFFAOYSA-N | |
7390 | |
CC1=CC=C(C=C1)C(C)(C)C |
C11H16 | |
MFCD00008837 | |
4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene | |
1-tert-butyl-4-methylbenzene |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Fatal if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a w
GHS Signal Word: Warning
EINECSNumber : 202-675-9
RUO – Research Use Only