Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butyltoluene, 95%

Chemical Identifiers

Chemical Name or Material 4-tert-Butyltoluene
CAS 98-51-1
Molecular Formula C11H16
Formula Weight 148.25
MDL Number MFCD00008837
UN Number UN2667
Beilstein 2038670
Synonym 4-tert-Butyl-1-methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA18214AP

  SDS
alfa aesar™
A18214AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.25
Assay 95%
Quantity 500mL
Health Hazard 3 GHS P Statement
P210-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2038670
TSCA TSCA
Chemical Name or Material 4-tert-Butyltoluene
UN Number UN2667
Boiling Point 189° to 193°C
RTECS Number XS8400000
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BUTYLTOLUENES
Synonym 4-tert-Butyl-1-methylbenzene
Health Hazard 1 Danger
MDL Number MFCD00008837
Health Hazard 2 GHS H Statement
H330-H227-H302-H319-H335
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point −63°C
CAS 98-51-1
Flash Point 62°C (144°F)
Molecular Formula C11H16
Odor Characteristic

Applications
4-tert-Butyltoluene is used as pharmaceutical intermediate.

Solubility
Soluble in water (6 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H227-H302-H319-H335
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.