Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylthiophenol gold nanoparticles, 2% (w/v) soln. in toluene, 2-5nm particles

Chemical Identifiers

Chemical Name or Material 4-tert-Butylthiophenol gold nanoparticles
Name Note 2 to 5nm particles
MDL Number MFCD09881474
UN Number UN1294
Synonym Au-tert-butylbenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH28235AA

 
alfa aesar™
H28235AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-tert-Butylthiophenol gold nanoparticles
Name Note 2 to 5nm particles
MDL Number MFCD09881474
UN Number UN1294
Synonym Au-tert-butylbenzenethiol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 111°C
Flash Point 4°C (39°F)
Odor Aromatic
Concentration 2% (w/v) soln. in toluene
Quantity 5mL
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE

Applications
The study of gold nanoparticles is very captivating because of their significance and applications as catalysts in restructuring technologies that are used for manufacturing, medicine, energy production, transportation, computation, communication, and environmental changes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.