Learn More
4-tert-Butylthiophenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1214814
Description
4-tert-butylthiophenol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-tert-butylthiophenol is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents.
Specifications
4-tert-Butylthiophenol | |
-11°C | |
238°C | |
Stench | |
1.5485 | |
25 g | |
Air Sensitive | |
Insoluble in water. | |
CC(C)(C)C1=CC=C(C=C1)S | |
166.282 | |
166.29 |
2396-68-1 | |
0.99 | |
>110°C (230°F) | |
C10H14S | |
MFCD00022067 | |
606476 | |
4-tert-butylthiophenol, p-tert-butylthiophenol, benzenethiol, p-tert-butyl, p-tert-butylbenzenethiol, p-t-butylthiophenol, p-tert-butylphenyl mercaptan, benzenethiol, 4-1,1-dimethylethyl, benzenethiol, p-tert-butylthio, 4-tert-butyl thiophenol, 4-t-butylthiophenol | |
GNXBFFHXJDZGEK-UHFFFAOYSA-N | |
4-tert-butylbenzenethiol | |
75454 | |
97% |
Chemical Identifiers
2396-68-1 | |
166.282 | |
GNXBFFHXJDZGEK-UHFFFAOYSA-N | |
75454 | |
CC(C)(C)C1=CC=C(C=C1)S |
C10H14S | |
MFCD00022067 | |
4-tert-butylthiophenol, p-tert-butylthiophenol, benzenethiol, p-tert-butyl, p-tert-butylbenzenethiol, p-t-butylthiophenol, p-tert-butylphenyl mercaptan, benzenethiol, 4-1,1-dimethylethyl, benzenethiol, p-tert-butylthio, 4-tert-butyl thiophenol, 4-t-butylthiophenol | |
4-tert-butylbenzenethiol |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-255-6
RTECSNumber : DC1025000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only