Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylthiobenzamide, 97%

$41.11 - $286.31

Chemical Identifiers

Chemical Name or Material 4-tert-Butylthiobenzamide
CAS 57774-77-3
Molecular Formula C11H15NS
Formula Weight 193.31
MDL Number MFCD00052485
Synonym 4-tert-Butylbenzenecarbothioamide; 4-(tert-Butyl)benzene-1-carbothioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2651903

  SDS
alfa aesar™
H2651903
1g Each for $95.74

AAH2651906

  SDS
alfa aesar™
H2651906
5g Each for $286.31

AAH26519MD

  SDS
alfa aesar™
H26519MD
250mg Each for $41.11
Description & Specifications

Specifications

Formula Weight 193.31
Chemical Name or Material 4-tert-Butylthiobenzamide
Assay 97%
Molecular Formula C11H15NS
MDL Number MFCD00052485
CAS 57774-77-3
Synonym 4-tert-Butylbenzenecarbothioamide; 4-(tert-Butyl)benzene-1-carbothioamide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 145° to 149°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.