missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylstyrene, 94%, stab. with 50 ppm 4-tert-butylcatechol

Manufacturer:  Alfa Aesar™ L12356AP

 View more versions of this product

Catalog No. AAL12356AP



Description

Description

Applications
4-tert-Butylstyrene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in the dark. Protect from heat. Incompatible with heat, light, free radical initiators, UV radiation. Do not store together.
Specifications

Specifications

4-tert-Butylstyrene
219°C
500mL
1746-23-2
MFCD00065126
4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
CC(C)(C)C1=CC=C(C=C1)C=C
160.26
160.26
80°C (176°F)
-38°C
stabilized with 50ppm 4-tert-butylcatechol
C12H16
1903714
QEDJMOONZLUIMC-UHFFFAOYSA-N
1-tert-butyl-4-ethenylbenzene
15627
94%

Chemical Identifiers

1746-23-2
160.26
QEDJMOONZLUIMC-UHFFFAOYSA-N
15627
CC(C)(C)C1=CC=C(C=C1)C=C
C12H16
MFCD00065126
4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
1-tert-butyl-4-ethenylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents