missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-tert-Butylpropiophenone, TCI America™

$131.39 - $392.64

Chemical Identifiers

CAS 71209-71-7
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.286
MDL Number MFCD00100434
InChI Key AQNYEAINONORRY-UHFFFAOYSA-N
Synonym 4-tert-Butylpropionylbenzene
PubChem CID 595281
IUPAC Name 1-(4-tert-butylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16091ML View Documents TCI America
B16091ML
1mL Each for $131.39
 
B16095ML View Documents TCI America
B16095ML
5mL Each for $392.64
 
Specifications

Chemical Identifiers

71209-71-7
190.286
AQNYEAINONORRY-UHFFFAOYSA-N
595281
CCC(=O)C1=CC=C(C=C1)C(C)(C)C
C13H18O
MFCD00100434
4-tert-Butylpropionylbenzene
1-(4-tert-butylphenyl)propan-1-one

Specifications

162°C
1mL
C13H18O
1993
AQNYEAINONORRY-UHFFFAOYSA-N
1-(4-tert-butylphenyl)propan-1-one
595281
Clear Liquid at 20°C
4′-tert-Butylpropiophenone
Yellow
71209-71-7
MFCD00100434
4-tert-Butylpropionylbenzene
CCC(=O)C1=CC=C(C=C1)C(C)(C)C
190.286
190.29
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No