missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylphthalonitrile, TCI America™

$215.42 - $727.23

Chemical Identifiers

CAS 32703-80-3
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00060112
InChI Key LOTMIRVNJTVTSU-UHFFFAOYSA-N
Synonym 4-tert-Butyl-1,2-dicyanobenzene
PubChem CID 122931
IUPAC Name 4-tert-butylbenzene-1,2-dicarbonitrile
SMILES CC(C)(C)C1=CC(=C(C=C1)C#N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B13345G View Documents TCI America
B13345G
5g Each for $215.42
 
B133425G View Documents TCI America
B133425G
25g Each for $727.23
 
Specifications

Chemical Identifiers

32703-80-3
184.242
LOTMIRVNJTVTSU-UHFFFAOYSA-N
122931
CC(C)(C)C1=CC(=C(C=C1)C#N)C#N
C12H12N2
MFCD00060112
4-tert-Butyl-1,2-dicyanobenzene
4-tert-butylbenzene-1,2-dicarbonitrile

Specifications

165°C
60°C
32703-80-3
MFCD00060112
LOTMIRVNJTVTSU-UHFFFAOYSA-N
4-tert-butylbenzene-1,2-dicarbonitrile
122931
Crystal-Powder at 20°C
4-tert-Butylphthalonitrile
White-Yellow
5g
C12H12N2
4-tert-Butyl-1,2-dicyanobenzene
CC(C)(C)C1=CC(=C(C=C1)C#N)C#N
184.242
184.24
≥98.0% (GC,N)
Safety and Handling

Safety and Handling

TSCA : No