Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylphenylhydrazine hydrochloride, 97%

$152.39 - $484.88

Chemical Identifiers

Chemical Name or Material 4-tert-Butylphenylhydrazine hydrochloride
CAS 128231-55-0
Molecular Formula C10H16N2·HCl
Formula Weight 200.71
MDL Number MFCD00013387
Beilstein 3700093
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1438506

  SDS
alfa aesar™
A1438506
5g Each for $152.39

AAA1438514

  SDS
alfa aesar™
A1438514
25g Each for $484.88
Description & Specifications

Specifications

Formula Weight 200.71
Chemical Name or Material 4-tert-Butylphenylhydrazine hydrochloride
Assay 97%
Molecular Formula C10H16N2·HCl
MDL Number MFCD00013387
CAS 128231-55-0
Beilstein 3700093
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point ≈228°C (dec.)
Quantity 5g

Applications
It is used in the ring transformations of heterocyclic compounds and in the identification, synthesis, and pharmacological evaluation of tetrahydroindazole based ligands.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.