missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylphenyl glycidyl ether, 95%

Manufacturer:  Alfa Aesar™ H5594506

 View more versions of this product

Catalog No. AAH5594506



Description

Description

Applications
4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

4-tert-Butylphenyl glycidyl ether
102°C (216°F)
165°C to 170°C (14mmHg)
3101-60-8
MFCD00005136
Slightly soluble in water.
CC(C)(C)C1=CC=C(C=C1)OCC2CO2
206.285
206.28
95%
1.038
1.515
5g
C13H18O2
2-4-tert-butyl phenoxy methyl oxirane
HHRACYLRBOUBKM-UHFFFAOYSA-N
2-[(4-tert-butylphenoxy)methyl]oxirane
18360
Liquid

Chemical Identifiers

3101-60-8
206.285
HHRACYLRBOUBKM-UHFFFAOYSA-N
18360
CC(C)(C)C1=CC=C(C=C1)OCC2CO2
C13H18O2
MFCD00005136
2-4-tert-butyl phenoxy methyl oxirane
2-[(4-tert-butylphenoxy)methyl]oxirane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 221-453-2

RTECSNumber : TX4250000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents