missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(tert-Butyldimethylsilyloxy)-1-butyne, TCI America™

$139.03 - $414.03

Chemical Identifiers

CAS 78592-82-2
Molecular Formula C10H20OSi
Molecular Weight (g/mol) 184.354
MDL Number MFCD03427046
InChI Key NUZBJLXXTAOBPH-UHFFFAOYSA-N
Synonym (But-3-yn-1-yloxy)(tert-butyl)dimethylsilane
PubChem CID 4304567
IUPAC Name tert-butyl-but-3-ynoxy-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B50421ML View Documents TCI America
B50421ML
1mL Each for $139.03
 
B50425ML View Documents TCI America
B50425ML
5mL Each for $414.03
 
Specifications

Chemical Identifiers

78592-82-2
184.354
NUZBJLXXTAOBPH-UHFFFAOYSA-N
4304567
CC(C)(C)[Si](C)(C)OCCC#C
C10H20OSi
MFCD03427046
(But-3-yn-1-yloxy)(tert-butyl)dimethylsilane
tert-butyl-but-3-ynoxy-dimethylsilane

Specifications

83°C
1mL
C10H20OSi
1993
NUZBJLXXTAOBPH-UHFFFAOYSA-N
tert-butyl-but-3-ynoxy-dimethylsilane
4304567
Clear Liquid at 20°C
4-(tert-Butyldimethylsilyloxy)-1-butyne
Yellow
78592-82-2
MFCD03427046
(But-3-yn-1-yloxy)(tert-butyl)dimethylsilane
CC(C)(C)[Si](C)(C)OCCC#C
184.354
184.35
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No