missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(tert-Butyldimethylsiloxy)phenol, 97%

Manufacturer:  Alfa Aesar™ H5555603

 View more versions of this product

Catalog No. AAH5555603



Specifications

Specifications

4-(tert-Butyldimethylsiloxy)phenol
60°C to 64°C
108534-47-0
MFCD06200713
KZTVHIZALLBXMO-UHFFFAOYSA-N
4-[tert-butyl(dimethyl)silyl]oxyphenol
597388
Powder
White
1g
C12H20O2Si
4-t-butyldimethylsiloxy phenol
CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)O
224.375
224.38
97%

Chemical Identifiers

108534-47-0
224.375
KZTVHIZALLBXMO-UHFFFAOYSA-N
597388
CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)O
C12H20O2Si
MFCD06200713
4-t-butyldimethylsiloxy phenol
4-[tert-butyl(dimethyl)silyl]oxyphenol
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents