Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(tert-Butyldimethylsiloxy)phenol, 97%

Chemical Identifiers

Chemical Name or Material 4-(tert-Butyldimethylsiloxy)phenol
CAS 108534-47-0
Formula Weight 224.38
MDL Number MFCD06200713
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5555603

 
alfa aesar™
H5555603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5555606

 
alfa aesar™
H5555606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55556MD

 
alfa aesar™
H55556MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.38
Melting Point 60° to 64°C
Chemical Name or Material 4-(tert-Butyldimethylsiloxy)phenol
Physical Form Powder
Assay 97%
MDL Number MFCD06200713
CAS 108534-47-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Color White to off-white
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.