Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butyldimethylsiloxy-3-penten-2-one, 95%, mixture of isomers

$92.81 - $372.50

Chemical Identifiers

Chemical Name or Material 4-tert-Butyldimethylsiloxy-3-penten-2-one
CAS 69404-97-3
Name Note mixture of isomers
Molecular Formula C11H22O2Si
Formula Weight 214.38
MDL Number MFCD00010655
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2786806

  SDS
alfa aesar™
H2786806
5g Each for $92.81

AAH2786814

  SDS
alfa aesar™
H2786814
25g Each for $372.50
Description & Specifications

Specifications

Formula Weight 214.38
Chemical Name or Material 4-tert-Butyldimethylsiloxy-3-penten-2-one
Name Note mixture of isomers
Assay 95%
Molecular Formula C11H22O2Si
MDL Number MFCD00010655
CAS 69404-97-3
Boiling Point 50° to 51°C (0.2mmHg)
Quantity 5g
Flash Point 73°C (163°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.