Please login to your online account to display your discounted pricing

4-tert-Butylcyclohexanol, Mixture of Isomers 99%, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  107902500

 View more versions of this product

Catalog No. AC107902500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Granules
Assay ≥98.5%
Formula Weight 156.27
Quantity 250g
Color White
Beilstein 06, I, 18
EINECS Number 202-676-4
TSCA TSCA
Melting Point 60° to 69°C (depending on cis/trans ratio)
RTECS Number GV8750000
CAS Max % 100.0
Chemical Name or Material 4-tert-Butylcyclohexanol
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H20O
Linear Formula (CH3)3CC6H10OH
MDL Number MFCD00001473
CAS 98-52-2
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Plastic bottle
Additional Information cis/trans ratio: approximately 70% trans, 30% cis

Specifications

Chemical Name or Material 4-tert-Butylcyclohexanol
Name Note 99%
CAS 98-52-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H20O
Linear Formula (CH3)3CC6H10OH
Formula Weight 156.27
MDL Number MFCD00001473