Learn More
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 154355000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-tert-Butylcatechol | |
98-29-3 | |
100.0 | |
1.0490g/mL | |
129°C | |
Plastic bottle | |
(CH3)3CC6H3(OH)2 | |
500 g | |
15,54 | |
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin | |
XESZUVZBAMCAEJ-UHFFFAOYSA-N | |
4-tert-butylbenzene-1,2-diol | |
7381 | |
166.22 | |
Technical |
99% | |
98.5 | |
53.0°C to 56.0°C | |
285.0°C | |
0.7% max. Catechol (GC) | |
C10H14O2 | |
MFCD00002201 | |
06,IV,6014 | |
1.049 | |
Solubility in water: 0.2% (25°C). Other solubilities: soluble in hot water,very sol. in alcohol,ethers,esters and ketones,partly sol. in chlorinated and aromatic hydrocarb.,slightly sol. in aliphatics. 240g/100g acetone,80g/100g benzene;60g/100g | |
CC(C)(C)C1=CC=C(O)C(O)=C1 | |
166.22 | |
90 mm2/s (25°C) | |
99% |
Chemical Identifiers
98-29-3 | |
166.22 | |
XESZUVZBAMCAEJ-UHFFFAOYSA-N | |
7381 | |
CC(C)(C)C1=CC=C(O)C(O)=C1 |
C10H14O2 | |
MFCD00002201 | |
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin | |
4-tert-butylbenzene-1,2-diol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting eff
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 202-653-9
RTECSNumber : UX1400000
TSCA : TSCA
RUO – Research Use Only