missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylcatechol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 154350050

 View more versions of this product

Catalog No. AC154350050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-tert-Butylcatechol
98-29-3
100.0
Technical Grade
99%
(CH3)3CC6H3(OH)2
MFCD00002201
99%
98.5
Plastic bottle
5g
C10H14O2
166.22
4-(1, 1-Dimethylethyl)-1, 2-benzenediol

Chemical Identifiers

98-29-3
100.0
C10H14O2
166.22
4-(1, 1-Dimethylethyl)-1, 2-benzenediol
Technical Grade
98.5
99%
(CH3)3CC6H3(OH)2
MFCD00002201
4-tert-Butylcatechol, 99%

Safety and Handling

SDS & Safety Documents

GHS P Statement Harmful if swallowed. Harmful in contact with skin. Causes severe skin burns and eye damage. Harmful to aquatic life with long lasting effects.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact

Danger

EINECSNumber : 202-653-9

RTECSNumber : UX1400000

TSCA : TSCA