missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(tert-Butylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5330106

 View more versions of this product

Catalog No. AAH5330106



Specifications

Specifications

4-(tert-Butylcarbamoyl)benzeneboronic acid
5g
C11H16BNO3
4-tert-butylcarbamoyl phenyl boronic acid, 4-tert-butylaminocarbonyl phenylboronic acid, 4-tert-butylcarbamoyl phenylboronic acid, 4-tert-butylcarbamoyl phenyl boronic acid, 4-tert-butylaminocarbonyl benzeneboronic acid, 4-tert-butylcarbamoyl benzeneboronic acid, n-t-butyl 4-boronobenzamide, n-tert-butyl 4-boronobenzamide, 4-1,1-dimethylethyl amino carbonyl phenyl boronic acid, acmc-209q1e
B(C1=CC=C(C=C1)C(=O)NC(C)(C)C)(O)O
221.063
221.06
230°C to 246°C
850568-14-8
MFCD03788413
JAIIZPWLCVMPFA-UHFFFAOYSA-N
[4-(tert-butylcarbamoyl)phenyl]boronic acid
2773312
97%

Chemical Identifiers

850568-14-8
221.063
JAIIZPWLCVMPFA-UHFFFAOYSA-N
2773312
B(C1=CC=C(C=C1)C(=O)NC(C)(C)C)(O)O
C11H16BNO3
MFCD03788413
4-tert-butylcarbamoyl phenyl boronic acid, 4-tert-butylaminocarbonyl phenylboronic acid, 4-tert-butylcarbamoyl phenylboronic acid, 4-tert-butylcarbamoyl phenyl boronic acid, 4-tert-butylaminocarbonyl benzeneboronic acid, 4-tert-butylcarbamoyl benzeneboronic acid, n-t-butyl 4-boronobenzamide, n-tert-butyl 4-boronobenzamide, 4-1,1-dimethylethyl amino carbonyl phenyl boronic acid, acmc-209q1e
[4-(tert-butylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents