missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylcalix[8]arene, TCI America™

$108.47 - $362.09

Chemical Identifiers

CAS 68971-82-4
Molecular Formula C88H112O8
Molecular Weight (g/mol) 1297.856
MDL Number MFCD00075392
InChI Key OLZFZIXORGGLLS-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 8 arene, 4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
PubChem CID 434172
SMILES CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17505G View Documents TCI America
B17505G
5g Each for $108.47
 
B175025G View Documents TCI America
B175025G
25g Each for $362.09
 
Specifications

Chemical Identifiers

68971-82-4
1297.856
OLZFZIXORGGLLS-UHFFFAOYSA-N
434172
C88H112O8
MFCD00075392
4-tert-butylcalix 8 arene, 4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O

Specifications

White
68971-82-4
MFCD00075392
OLZFZIXORGGLLS-UHFFFAOYSA-N
1297.856
1297.86
≥98.0% (HPLC)
5g
C88H112O8
4-tert-butylcalix 8 arene, 4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O
434172
Crystal-Powder at 20°C
4-tert-Butylcalix[8]arene
Safety and Handling

Safety and Handling

TSCA : Yes