missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylcalix[8]arene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 290240250

 View more versions of this product

Catalog No. AC290240250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1297.87
≥96.0%
White to off-white
96.0
Complies
68971-82-4
C88H112O8
97%
Micro-Crystalline Powder
25g
100.0
Glass bottle
MFCD00075392
97%
4-tert-Butylcalix[8]arene
Authentic

Chemical Identifiers

68971-82-4
100.0
C88H112O8
MFCD00075392
≥96.0%
96.0
97%
1297.87
4-tert-Butylcalix[8]arene, 97%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA