missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-tert-Butylcalix[8]arene, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 290240025
Description
Chelation/Complexation reagentSpecifications
4-tert-Butylcalix[8]arene | |
Complies | |
White | |
2.5g | |
68971-82-4 | |
100.0 | |
C88H112O8 | |
3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, octa-tert-butyl octahydroxy calix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene | |
OLZFZIXORGGLLS-UHFFFAOYSA-N | |
1297.87 | |
1297.87 | |
97% |
Authentic | |
Glass bottle | |
411°C to 412°C | |
97% | |
96.0 | |
96% min. (HPLC) | |
MFCD00075392 | |
Solubility in water: insoluble in water. Other solubilities: soluble in chcl3,ch2cl2 and toluene | |
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O | |
434172 | |
Micro-Crystalline Powder |
Chemical Identifiers
68971-82-4 | |
1297.87 | |
OLZFZIXORGGLLS-UHFFFAOYSA-N | |
434172 |
C88H112O8 | |
MFCD00075392 | |
3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, octa-tert-butyl octahydroxy calix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene | |
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O |
Safety and Handling
TSCA : TSCA