missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
4-tert-Butylcalix[8]arene, 97%, ACROS Organics™
Chemical Identifiers
CAS |
68971-82-4 |
Molecular Formula |
C88H112O8 |
Molecular Weight (g/mol) |
1297.87 |
MDL Number |
MFCD00075392 |
InChI Key |
OLZFZIXORGGLLS-UHFFFAOYSA-N |
Synonym |
3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, octa-tert-butyl octahydroxy calix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene |
PubChem CID |
434172 |
SMILES |
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
AC290240025
SDS
View Documents
|
Thermo Fisher Scientific
290240025
|
2.5g |
Glass bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
Description
Description
Chelation/Complexation reagent
Specifications
Chemical Identifiers
|
68971-82-4 |
|
1297.87 |
|
OLZFZIXORGGLLS-UHFFFAOYSA-N |
|
434172 |
|
C88H112O8 |
|
MFCD00075392 |
|
3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, octa-tert-butyl octahydroxy calix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene |
|
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O |
Specifications
|
Authentic |
|
Glass bottle |
|
411°C to 412°C |
|
68971-82-4 |
|
100.0 |
|
C88H112O8 |
|
3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, 4-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, octa-tert-butyl octahydroxy calix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene |
|
OLZFZIXORGGLLS-UHFFFAOYSA-N |
|
1297.87 |
|
1297.87 |
|
97% |
|
Complies |
|
White |
|
2.5g |
|
96.0 |
|
96% min. (HPLC) |
|
MFCD00075392 |
|
Solubility in water: insoluble in water. Other solubilities: soluble in chcl3,ch2cl2 and toluene |
|
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)C)CC8=C(C(=CC(=C8)C(C)(C)C)CC9=C(C(=CC(=C9)C(C)(C)C)C2)O)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)O |
|
434172 |
|
Micro-Crystalline Powder |
|
4-tert-Butylcalix[8]arene, 97% |
Safety and Handling
SDS