missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylbiphenyl, TCI America™

$158.89 - $476.67

Chemical Identifiers

CAS 1625-92-9
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00222366
InChI Key CDOYZTOFTGTGBC-UHFFFAOYSA-N
PubChem CID 137120
IUPAC Name 1-tert-butyl-4-phenylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18115G View Documents TCI America
B18115G
5g Each for $158.89
 
B181125G View Documents TCI America
B181125G
25g Each for $476.67
 
Specifications

Chemical Identifiers

1625-92-9
210.32
CDOYZTOFTGTGBC-UHFFFAOYSA-N
1-tert-butyl-4-phenylbenzene
C16H18
MFCD00222366
137120
CC(C)(C)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

310°C
52°C
1625-92-9
MFCD00222366
CC(C)(C)C1=CC=C(C=C1)C2=CC=CC=C2
210.32
210.32
≥98.0% (GC)
White
5g
C16H18
CDOYZTOFTGTGBC-UHFFFAOYSA-N
1-tert-butyl-4-phenylbenzene
137120
Crystal-Powder at 20°C
4-tert-Butylbiphenyl
Safety and Handling

Safety and Handling

TSCA : No