missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylbenzonitrile, TCI America™

$111.53 - $369.73

Chemical Identifiers

CAS 4210-32-6
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00075840
InChI Key IIZURLNRIMKEDL-UHFFFAOYSA-N
Synonym 4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
PubChem CID 77883
IUPAC Name 4-tert-butylbenzonitrile
SMILES CC(C)(C)C1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B15765ML View Documents TCI America
B15765ML
5mL Each for $111.53
 
B157625ML View Documents TCI America
B157625ML
25mL Each for $369.73
 
Specifications

Chemical Identifiers

4210-32-6
159.232
IIZURLNRIMKEDL-UHFFFAOYSA-N
77883
CC(C)(C)C1=CC=C(C=C1)C#N
C11H13N
MFCD00075840
4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
4-tert-butylbenzonitrile

Specifications

112°C
5mL
C11H13N
3276
IIZURLNRIMKEDL-UHFFFAOYSA-N
4-tert-butylbenzonitrile
77883
Clear Liquid at 20°C
4-tert-Butylbenzonitrile
Yellow
4210-32-6
MFCD00075840
4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
CC(C)(C)C1=CC=C(C=C1)C#N
159.232
159.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No