Please login to your online account to display your discounted pricing

4-tert-Butylbenzoic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-tert-Butylbenzoic acid
CAS 98-73-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11 H14 O2
Linear Formula (CH3)3CC6H4CO2H
Formula Weight 178.23
MDL Number MFCD00002563
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC165632500

  SDS
thermo fisher scientific
AC165632500
Plastic bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Flakes
Assay ≥98.5%
Quantity 250g
Color White to yellow
Beilstein 09, 560
Chemical Name or Material 4-tert-Butylbenzoic acid
MDL Number MFCD00002563
CAS 98-73-7
Synonym PTBBA
Linear Formula (CH3)3CC6H4CO2H
CAS Max % 100.0
CAS Min % 98.5
Molecular Formula C11 H14 O2
Packaging Plastic bottle
Assay Percent Range 99%
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement May damage fertility. Causes damage to organs through prolonged or repeated exposure if inhaled. Harmful if swallowed.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Use person
Melting Point 164° to 167°C
Formula Weight 178.23
Safety and Handling

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Use person

GHS P Statement May damage fertility. Causes damage to organs through prolonged or repeated exposure if inhaled. Harmful if swallowed.