Please login to your online account to display your discounted pricing

4-tert-Butylbenzoic acid, 99%, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC165632500

 View more versions of this product

Catalog No. AC165632500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Flakes
Assay ≥98.5%
Quantity 250g
Color White to yellow
Beilstein 09, 560
Chemical Name or Material 4-tert-Butylbenzoic acid
MDL Number MFCD00002563
CAS 98-73-7
Synonym PTBBA
Linear Formula (CH3)3CC6H4CO2H
CAS Max % 100.0
CAS Min % 98.5
Molecular Formula C11 H14 O2
Packaging Plastic bottle
Assay Percent Range 99%
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement May damage fertility. Causes damage to organs through prolonged or repeated exposure if inhaled. Harmful if swallowed.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Use person
Melting Point 164° to 167°C
Formula Weight 178.23

Specifications

Chemical Name or Material 4-tert-Butylbenzoic acid
CAS 98-73-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11 H14 O2
Linear Formula (CH3)3CC6H4CO2H
Formula Weight 178.23
MDL Number MFCD00002563
Synonym PTBBA
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Use person

GHS P Statement May damage fertility. Causes damage to organs through prolonged or repeated exposure if inhaled. Harmful if swallowed.

SDS & Safety Documents