missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylbenzenesulfonyl chloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC303470500

 View more versions of this product

Catalog No. AC303470500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-tert-Butylbenzenesulfonyl chloride
97.5
Authentic
≥97.5% (GC)
Glass bottle
(CH3)3CC6H4SOCl
MFCD00007449
78° to 84°C
15084-51-2
100.0
Crystalline Powder
98%
C10H13ClO2S
232.73
White to light yellow
50g

Chemical Identifiers

15084-51-2
100.0
C10H13ClO2S
232.73
4-tert-Butylbenzenesulfonyl chloride
97.5
98%
(CH3)3CC6H4SOCl
MFCD00007449
≥97.5% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger