Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylbenzamidoxime, 97%

$295.66 - $295.66

Chemical Identifiers

Chemical Name or Material 4-tert-Butylbenzamidoxime
CAS 175204-39-4
Molecular Formula C11H16N2O
Formula Weight 192.26g/mol
MDL Number MFCD00082855
Synonym 4-tert-Butyl-N-hydroxybenzenecarboximidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5065206

 
alfa aesar™
H50652-06
Each for $295.66
Description & Specifications

Specifications

Melting Point 139° to 141°C
Quantity 5g
Chemical Name or Material 4-tert-Butylbenzamidoxime
CAS 175204-39-4
Assay 97%
Molecular Formula C11H16N2O
Formula Weight 192.26g/mol
MDL Number MFCD00082855
Synonym 4-tert-Butyl-N-hydroxybenzenecarboximidamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.