missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylbenzamide, 98%

Manufacturer:  Alfa Aesar™ A1538030

 View more versions of this product

Catalog No. AAA1538030



Specifications

Specifications

4-tert-Butylbenzamide
250g
C11H15NO
2086692
177.25
171°C to 175°C
56108-12-4
MFCD00017139
4-tert-butyl benzamide, benzamide, 4-1,1-dimethylethyl, p-tert-butyl benzamide, 4-tert-butyl benzenecarboxamide, unii-5e7907z6bm, 4-1,1-dimethylethyl benzamide, p-t-butylbenzamide, 4-t-butylbenzamide, acmc-20amap, butylbenzenecarboxamide
98%

Chemical Identifiers

56108-12-4
C11H15NO
MFCD00017139
4-tert-butyl benzamide, benzamide, 4-1,1-dimethylethyl, p-tert-butyl benzamide, 4-tert-butyl benzenecarboxamide, unii-5e7907z6bm, 4-1,1-dimethylethyl benzamide, p-t-butylbenzamide, 4-t-butylbenzamide, acmc-20amap, butylbenzenecarboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents