missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylbenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ A1574006

 View more versions of this product

Catalog No. AAA1574006



Description

Description

Applications
Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.

Solubility
Soluble in water.

Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Specifications

Specifications

4-tert-Butylbenzaldehyde
244°C to 246°C
939-97-9
MFCD00035742
1906461
OTXINXDGSUFPNU-UHFFFAOYSA-N
4-tert-butylbenzaldehyde
70324
97%
101°C (213°F)
5g
C11H14O
UN3082
4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
CC(C)(C)C1=CC=C(C=C1)C=O
162.232
162.23

Chemical Identifiers

939-97-9
162.232
OTXINXDGSUFPNU-UHFFFAOYSA-N
70324
CC(C)(C)C1=CC=C(C=C1)C=O
C11H14O
MFCD00035742
4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
4-tert-butylbenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA

Documents