Please login to your online account to display your discounted pricing

4-tert-Butylbenzaldehyde, 95%, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426522500

 View more versions of this product

Catalog No. AC426522500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 162.23
Quantity 250mL
Color Clear colorless to yellow
CAS Min % 94.0
CAS Max % 100.0
Linear Formula (CH3)3CC6H4CHO
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-tert-Butylbenzaldehyde
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C11H14O
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
MDL Number MFCD00035742
CAS 939-97-9

Specifications

Chemical Name or Material 4-tert-Butylbenzaldehyde
CAS 939-97-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H14O
Linear Formula (CH3)3CC6H4CHO
Formula Weight 162.23
MDL Number MFCD00035742
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.