Please login to your online account to display your discounted pricing

4-tert-Butylbenzaldehyde, 95%, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426520010

 View more versions of this product

Catalog No. AC426520010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 162.23
Physical Form Liquid
Assay ≥94%
Quantity 1L
Color Clear colorless to yellow
Density 0.9700g/mL
CAS Min % 94.0
CAS Max % 100.0
Linear Formula (CH3)3CC6H4CHO
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Chemical Name or Material 4-tert-Butylbenzaldehyde
Assay Percent Range 95%
Molecular Formula C11H14O
MDL Number MFCD00035742
CAS 939-97-9
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-tert-Butylbenzaldehyde
CAS 939-97-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H14O
Linear Formula (CH3)3CC6H4CHO
Formula Weight 162.23
MDL Number MFCD00035742
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.