missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylanisole, TCI America™

$113.06 - $945.72

Chemical Identifiers

CAS 5396-38-3
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00026309
InChI Key MCUPBIBNSTXCPQ-UHFFFAOYSA-N
Synonym 4-tert-butylanisole, 1-tert-butyl-4-methoxybenzene, p-tert-butylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, 4-tert-butylphenyl methyl ether, 1-tert-butyl-4-methoxy-benzene, 4-t-butylanisole, 4-tertbutylanisole
PubChem CID 94750
IUPAC Name 1-tert-butyl-4-methoxybenzene
SMILES CC(C)(C)C1=CC=C(C=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B096125ML View Documents TCI America
B096125ML
25mL Each for $113.06
 
B0961500ML View Documents TCI America
B0961500ML
500mL Each for $945.72
 
Specifications

Chemical Identifiers

5396-38-3
164.248
MCUPBIBNSTXCPQ-UHFFFAOYSA-N
94750
CC(C)(C)C1=CC=C(C=C1)OC
C11H16O
MFCD00026309
4-tert-butylanisole, 1-tert-butyl-4-methoxybenzene, p-tert-butylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, 4-tert-butylphenyl methyl ether, 1-tert-butyl-4-methoxy-benzene, 4-t-butylanisole, 4-tertbutylanisole
1-tert-butyl-4-methoxybenzene

Specifications

222°C
25mL
C11H16O
4-tert-butylanisole, 1-tert-butyl-4-methoxybenzene, p-tert-butylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, 4-tert-butylphenyl methyl ether, 1-tert-butyl-4-methoxy-benzene, 4-t-butylanisole, 4-tertbutylanisole
CC(C)(C)C1=CC=C(C=C1)OC
164.248
164.25
≥98.0% (GC)
Yellow
5396-38-3
MFCD00026309
MCUPBIBNSTXCPQ-UHFFFAOYSA-N
1-tert-butyl-4-methoxybenzene
94750
Clear Liquid at 20°C
4-tert-Butylanisole
Safety and Handling

Safety and Handling

TSCA : Yes