missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylanisole 98.0+%, TCI America™

Manufacturer:  TCI America B096125ML

 View more versions of this product

Catalog No. B096125ML



Specifications

Specifications

4-tert-Butylanisole
Yellow
5396-38-3
MFCD00026309
MCUPBIBNSTXCPQ-UHFFFAOYSA-N
1-tert-butyl-4-methoxybenzene
94750
Clear Liquid at 20°C
222°C
25mL
C11H16O
4-tert-butylanisole, 1-tert-butyl-4-methoxybenzene, p-tert-butylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, 4-tert-butylphenyl methyl ether, 1-tert-butyl-4-methoxy-benzene, 4-t-butylanisole, 4-tertbutylanisole
CC(C)(C)C1=CC=C(C=C1)OC
164.248
164.25
≥98.0% (GC)

Chemical Identifiers

5396-38-3
164.248
MCUPBIBNSTXCPQ-UHFFFAOYSA-N
94750
CC(C)(C)C1=CC=C(C=C1)OC
C11H16O
MFCD00026309
4-tert-butylanisole, 1-tert-butyl-4-methoxybenzene, p-tert-butylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, 4-tert-butylphenyl methyl ether, 1-tert-butyl-4-methoxy-benzene, 4-t-butylanisole, 4-tertbutylanisole
1-tert-butyl-4-methoxybenzene
Safety and Handling

Safety and Handling

TSCA : Yes

Documents