Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butylanisole, 98%

$41.43 - $468.61

Chemical Identifiers

Chemical Name or Material 4-tert-Butylanisole
CAS 5396-38-3
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00026309
Beilstein 2041952
Synonym 4-Methoxy-tert-butylbenzene; 4-Methoxy-tert-butylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2535706

  SDS
alfa aesar™
B2535706
5g Each for $41.43

AAB2535714

  SDS
alfa aesar™
B2535714
25g Each for $155.20

AAB2535722

  SDS
alfa aesar™
B2535722
100g Each for $468.61
Description & Specifications

Specifications

Formula Weight 164.25
Molecular Formula C11H16O
Flash Point 94°C (201°F)
TSCA TSCA
MDL Number MFCD00026309
Synonym 4-Methoxy-tert-butylbenzene; 4-Methoxy-tert-butylbenzene
Melting Point 18°C
Quantity 5g
Assay 98%
CAS 5396-38-3
Beilstein 2041952
Boiling Point 222°C
Chemical Name or Material 4-tert-Butylanisole