missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylanisole, 97%, ACROS Organics™

Chemical Identifiers

CAS 5396-38-3
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00026309
InChI Key MCUPBIBNSTXCPQ-UHFFFAOYSA-N
Synonym 1-tert-butyl-4-methoxy-benzene
PubChem CID 94750
IUPAC Name 1-tert-butyl-4-methoxybenzene
SMILES CC(C)(C)C1=CC=C(C=C1)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC371310250 SDS View Documents Thermo Fisher Scientific
AC371310250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

5396-38-3
164.25
MCUPBIBNSTXCPQ-UHFFFAOYSA-N
94750
CC(C)(C)C1=CC=C(C=C1)OC
C11H16O
MFCD00026309
1-tert-butyl-4-methoxy-benzene
1-tert-butyl-4-methoxybenzene

Specifications

0.9380g/mL
Authentic
1.5020 to 1.5040
Colorless to Yellow
5396-38-3
100.0
C11H16O
MFCD00026309
MCUPBIBNSTXCPQ-UHFFFAOYSA-N
1-tert-butyl-4-methoxybenzene
94750
Liquid
4-tert-Butylanisole
94°C
Glass bottle
0.938
25g
96.0
96% min. (GC)
(CH3)3CC6H4OCH3
1-tert-butyl-4-methoxy-benzene
CC(C)(C)C1=CC=C(C=C1)OC
164.25
164.25
97%
Safety and Handling

Safety and Handling

EINECSNumber : 226-412-2