missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-tert-Butylanisole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC371310250
Specifications
4-tert-Butylanisole | |
94°C | |
Glass bottle | |
0.938 | |
25g | |
96.0 | |
96% min. (GC) | |
(CH3)3CC6H4OCH3 | |
1-tert-butyl-4-methoxy-benzene, 1-tert-butyl-4-methoxybenzene, 4-t-butylanisole, 4-tert-butylanisole, 4-tert-butylphenyl methyl ether, 4-tertbutylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, p-tert-butylanisole | |
CC(C)(C)C1=CC=C(C=C1)OC | |
164.25 | |
164.25 | |
97% |
0.9380g/mL | |
Authentic | |
1.5020 to 1.5040 | |
Colorless to Yellow | |
5396-38-3 | |
100.0 | |
C11H16O | |
MFCD00026309 | |
MCUPBIBNSTXCPQ-UHFFFAOYSA-N | |
1-tert-butyl-4-methoxybenzene | |
94750 | |
Liquid |
Chemical Identifiers
5396-38-3 | |
164.25 | |
MCUPBIBNSTXCPQ-UHFFFAOYSA-N | |
94750 | |
CC(C)(C)C1=CC=C(C=C1)OC |
C11H16O | |
MFCD00026309 | |
1-tert-butyl-4-methoxy-benzene, 1-tert-butyl-4-methoxybenzene, 4-t-butylanisole, 4-tert-butylanisole, 4-tert-butylphenyl methyl ether, 4-tertbutylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, p-tert-butylanisole | |
1-tert-butyl-4-methoxybenzene |
Safety and Handling
EINECSNumber : 226-412-2