missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylaniline, 99%, ACROS Organics™

$201.71 - $201.71

Chemical Identifiers

CAS 769-92-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00007899
InChI Key WRDWWAVNELMWAM-UHFFFAOYSA-N
Synonym 4-t-butylaniline, 4-t-butylbenzeneamine, 4-tert-butyl aniline, 4-tert-butylbenzenamine, 4-tert-butylbenzeneamine, benzenamine, 4-1,1-dimethylethyl, p-t-butylaniline, p-tert-butylaniline, unii-47h9440zow
PubChem CID 69861
IUPAC Name 4-tert-butylaniline
SMILES CC(C)(C)C1=CC=C(C=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC198250050
SDS View Documents
Thermo Fisher Scientific
198250050
5mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC198250250
SDS View Documents
Thermo Fisher Scientific
198250250
25mL
Each for $201.71
 
Description

Description

Specifications

Chemical Identifiers

769-92-6
149.24
WRDWWAVNELMWAM-UHFFFAOYSA-N
69861
CC(C)(C)C1=CC=C(C=C1)N
C10H15N
MFCD00007899
4-t-butylaniline, 4-t-butylbenzeneamine, 4-tert-butyl aniline, 4-tert-butylbenzenamine, 4-tert-butylbenzeneamine, benzenamine, 4-1,1-dimethylethyl, p-t-butylaniline, p-tert-butylaniline, unii-47h9440zow
4-tert-butylaniline

Specifications

0.9300g/mL
Authentic
1.5370 to 1.539
230°C
15°C to 16°C
769-92-6
100.0
C10H15N
MFCD00007899
4-t-butylaniline, 4-t-butylbenzeneamine, 4-tert-butyl aniline, 4-tert-butylbenzenamine, 4-tert-butylbenzeneamine, benzenamine, 4-1,1-dimethylethyl, p-t-butylaniline, p-tert-butylaniline, unii-47h9440zow
CC(C)(C)C1=CC=C(C=C1)N
149.24
149.24
99%
101°C
Glass bottle
0.93
Brown to Orange-Red
5mL
98.5
98.5% min. (GC)
(CH3)3CC6H4NH2
12, 1166
WRDWWAVNELMWAM-UHFFFAOYSA-N
4-tert-butylaniline
69861
Liquid
4-tert-Butylaniline, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 212-215-9

SDS
Product Certifications