missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-tert-Butylacetophenone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 308740050

 View more versions of this product

Catalog No. AC308740050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4'-tert-Butylacetophenone
943-27-1
100.0
Liquid
98%
C12H16O
(CH3)3CC6H4COCH3
MFCD00017256
5g
98%
97.5
Authentic
≥97.5%
Glass bottle
1.5190 to 1.5220 (20°C, 589nm)
176.26
Clear colorless

Chemical Identifiers

943-27-1
100.0
C12H16O
176.26
4'-tert-Butylacetophenone, 98%
97.5
98%
(CH3)3CC6H4COCH3
MFCD00017256
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-399-3

TSCA : TSCA