Learn More
4-(tert-Butyl)benzylbromide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 345500050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(tert-Butyl)benzylbromide | |
96.0 | |
8°C to 12°C | |
1.2360g/mL | |
>109°C | |
96% min. (Argentometry) | |
C11H15Br | |
(CH3)3CC6H4CH2Br | |
5 g | |
4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide | |
CC(C)(C)C1=CC=C(C=C1)CBr | |
227.14 | |
227.14 | |
Liquid |
18880-00-7 | |
100.0 | |
Yellow | |
90°C to 94°C (1.5 mmHg) | |
Authentic | |
Glass bottle | |
1.5445 to 1.5465 | |
MFCD00000180 | |
1.236 | |
QZNQSIHCDAGZIA-UHFFFAOYSA-N | |
1-(bromomethyl)-4-tert-butylbenzene | |
87836 | |
97% |
Chemical Identifiers
18880-00-7 | |
227.14 | |
QZNQSIHCDAGZIA-UHFFFAOYSA-N | |
87836 | |
CC(C)(C)C1=CC=C(C=C1)CBr |
C11H15Br | |
MFCD00000180 | |
4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide | |
1-(bromomethyl)-4-tert-butylbenzene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 242-643-1
RUO – Research Use Only