Please login to your online account to display your discounted pricing

4-(tert-Butyl)benzene-1-carbothioamide, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(tert-Butyl)benzene-1-carbothioamide
CAS 57774-77-3
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H15NS
Linear Formula (CH3)3CC6H4CSNH2
Formula Weight 193.31
MDL Number MFCD00052485
Synonym 4-(tert-Butyl)benzene-1-carbothioamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF00754EA

  SDS
fisher scientific company
RF00754EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.31
Quantity 10g
Assay 95%
CAS Min % 95.0
Linear Formula (CH3)3CC6H4CSNH2
Synonym 4-(tert-Butyl)benzene-1-carbothioamide
CAS 57774-77-3
Assay Percent Range 95%
Molecular Formula C11H15NS
Chemical Name or Material 4-(tert-Butyl)benzene-1-carbothioamide
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00052485
Packaging Amber glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.